Order Entry
Germany
ContactUsLinkComponent
tert-Butyl-4-chloro-3-formyl-1H-indole-1-carboxylate 98%
tert-Butyl-4-chloro-3-formyl-1H-indole-1-carboxylate 98%
Catalog # APOSOR1684-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl-4-chloro-3-formyl-1H-indole-1-carboxylate 98%
Catalog # APOSOR1684-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :GHS07
Storage Temperature:Ambient
CAS Number:914349-00-1
Formula:C₁₄H₁₄ClNO₃