Order Entry
Germany
ContactUsLinkComponent
tert-Butyl-5-(benzyloxy)-3-formyl-1H-indole-1-carboxylate 98%
tert-Butyl-5-(benzyloxy)-3-formyl-1H-indole-1-carboxylate 98%
Catalog # APOSOR1674-250MG
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl-5-(benzyloxy)-3-formyl-1H-indole-1-carboxylate 98%
Catalog # APOSOR1674-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :GHS07
Storage Temperature:Ambient
CAS Number:914348-98-4
Formula:C₂₁H₂₁NO₄