4-Oxohexahydrocyclopenta[c]pyrrole-2-carboxylic acid tert-butyl ester 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:879686-42-7
:
Formula:C₁₂H₁₉NO₃
MDL Number:MFCD06658358
:225.29 g/mol
:GHS07
Storage Temperature:Ambient