Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(Furan-3-yl)ethanone
1-(Furan-3-yl)ethanone 97%
1-(Furan-3-yl)ethanone 97%
Irritant
1-(Furan-3-yl)ethanone 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1-(Furan-3-yl)ethanone 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
14313-09-8
:
Formula:
C₆H₆O₂
MDL Number:
MFCD01568491
:
110.112 g/mol
:
GHS07
Storage Temperature:
Ambient