Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(2R,3R)-(-)-2,3-Butanediol
(2R,3R)-(-)-2,3-Butanediol 99%
(2R,3R)-(-)-2,3-Butanediol 99%
:
Apollo Scientific
(2R,3R)-(-)-2,3-Butanediol 99%
:
Apollo Scientific
:
24347-58-8
:
:
C₄H₁₀O₂
:
MFCD00064267
:
90.12 g/mol
:
179…180 °C
:
0.987 g/cm³ (25 °C)
:
85 °C
:
246-186-9
:
GHS07
:
Ambient