Order Entry
Germany
ContactUsLinkComponent
N-Methyldopamine hydrochloride 98%
Irritant
N-Methyldopamine hydrochloride 98%
Catalog # APOSOR1575-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
N-Methyldopamine hydrochloride 98%
Catalog # APOSOR1575-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :203.67 g/mol
Melting Pt:180…181 °C
EINECS:200-528-3
 :GHS07
Storage Temperature:Ambient
CAS Number:62-32-8
Formula:C₉H₁₃NO₂·HCl
MDL Number:MFCD00035074