Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(R)-3-(Boc-amino)piperidine
(R)-3-(Boc-amino)piperidine
(R)-3-(Boc-amino)piperidine
(R)-3-(Boc-amino)piperidine
Supplier:
Apollo Scientific
Certificates
Expand image
(R)-3-(Boc-amino)piperidine
Supplier:
Apollo Scientific
About this item
CAS Number:
309956-78-3
:
Formula:
C₁₀H₂₀N₂O₂
MDL Number:
MFCD03093382
:
200.28 g/mol
:
GHS07
Storage Temperature:
Ambient