Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-2-chlorooxazole-4-carboxylate
Methyl-2-chlorooxazole-4-carboxylate 98%
Methyl-2-chlorooxazole-4-carboxylate 98%
Irritant
Methyl-2-chlorooxazole-4-carboxylate 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl-2-chlorooxazole-4-carboxylate 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
934236-35-8
:
Formula:
C₅H₄ClNO₃
MDL Number:
MFCD09265505
:
161.55 g/mol
:
GHS07
Storage Temperature:
Ambient