Order Entry
Germany
ContactUsLinkComponent
Methyl-3-phenyl-1H-pyrazole-5-carboxylate
Irritant
Methyl-3-phenyl-1H-pyrazole-5-carboxylate
Catalog # APOSOR15423-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-3-phenyl-1H-pyrazole-5-carboxylate
Catalog # APOSOR15423-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :202.21 g/mol
Melting Pt:180…182 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:856061-37-5
 :MFCD00208415
Formula:C₁₁H₁₀N₂O₂