4'-Propylacetophenone
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 1-(4-Propylphenyl)ethanone, p-Propylacetophenone
- CAS Number:2932-65-2
- Formula:C₁₁H₁₄O
- MDL Number:MFCD00041359
- Molecular Weight:162.23
- Boiling Pt:123…125 °C/15mmHg
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Synonyms: 1-(4-Propylphenyl)ethanone, p-Propylacetophenone
- CAS Number:2932-65-2
- Formula:C₁₁H₁₄O
- MDL Number:MFCD00041359
- Molecular Weight:162.23
- Boiling Pt:123…125 °C/15mmHg
- Storage Temperature:Ambient



