Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-4-piperidinobenzenecarboxylate
Methyl-4-piperidinobenzenecarboxylate
Methyl-4-piperidinobenzenecarboxylate
:
Apollo Scientific
Methyl-4-piperidinobenzenecarboxylate
:
Apollo Scientific
:
10338-58-6
:
:
C₁₃H₁₇NO₂
:
MFCD08444024
:
219.29 g/mol
:
94…96 °C
:
GHS07
:
Ambient