

2-Iodothiophene 98% stabilised
Catalog # APOSOR15030-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:210.04 g/mol
Boiling Pt:182 °C
Melting Pt:–40 °C
Density:2.064 g/cm³ (25 °C)
Flash Pt:71 °C
:Stabilised with copper
EINECS:222-342-1
:GHS05
:GHS07
Storage Temperature:Refrigerator
CAS Number:3437-95-4
Formula:C₄H₃IS
MDL Number:MFCD00005424