0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
1-Methyl-5-indoleboronic acid
1-Methyl-5-indoleboronic acid
1-Methyl-5-indoleboronic acid
Irritant
1-Methyl-5-indoleboronic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1-Methyl-5-indoleboronic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
192182-55-1
:
Formula:
C₉H₁₀BNO₂
MDL Number:
MFCD03412070
:
174.9900 g/mol
:
GHS07
Storage Temperature:
Freezer