0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
4-Methoxy-N-methyl-2-nitrobenzenamine
4-Methoxy-N-methyl-2-nitrobenzenamine 98%
4-Methoxy-N-methyl-2-nitrobenzenamine 98%
4-Methoxy-N-methyl-2-nitrobenzenamine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
4-Methoxy-N-methyl-2-nitrobenzenamine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
3360-79-0
:
Formula:
C₈H₁₀N₂O₃
MDL Number:
MFCD00962666
:
182.1800 g/mol
Melting Pt:
98…100 °C
:
GHS07
Storage Temperature:
Ambient