Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Methoxy-2-nitrobenzaldehyde
5-Methoxy-2-nitrobenzaldehyde 98%
5-Methoxy-2-nitrobenzaldehyde 98%
Irritant
5-Methoxy-2-nitrobenzaldehyde 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Methoxy-2-nitrobenzaldehyde 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
20357-24-8
:
Formula:
C₈H₇NO₄
MDL Number:
MFCD00456498
Melting Pt:
80…82 °C
:
181.1500 g/mol
:
GHS07
Storage Temperature:
Ambient