Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Boc-D-propargylglycine
Boc-D-propargylglycine 97%
Boc-D-propargylglycine 97%
Irritant
Boc-D-propargylglycine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Boc-D-propargylglycine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
63039-46-3
:
Formula:
C₁₀H₁₅NO₄
MDL Number:
MFCD01320886
:
213.233 g/mol
:
GHS07
Storage Temperature:
Ambient