0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Fmoc-Thi-OH
Fmoc-Thi-OH
Fmoc-Thi-OH
Fmoc-Thi-OH
Supplier:
Apollo Scientific
Certificates
Expand image
Fmoc-Thi-OH
Supplier:
Apollo Scientific
About this item
CAS Number:
130309-35-2
:
Formula:
C₂₂H₁₉NO₄S
MDL Number:
MFCD00065676
:
393.47 g/mol
:
GHS07
Storage Temperature:
Ambient