
N-Hydroxy-4-methoxybenzenecarboximidoyl chloride
Catalog # APOSOR14213-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:185.61 g/mol
Melting Pt:84…89 °C
:GHS07
Storage Temperature:Ambient
CAS Number:38435-51-7
Formula:C₈H₈ClNO₂
MDL Number:MFCD01924588