Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Hydrazino-1H-1,3-benzimidazole
2-Hydrazino-1H-1,3-benzimidazole
2-Hydrazino-1H-1,3-benzimidazole
:
Apollo Scientific
2-Hydrazino-1H-1,3-benzimidazole
:
Apollo Scientific
:
15108-18-6
:
:
C₇H₈N₄
:
MFCD00513603
:
148.17 g/mol
:
224…225 °C
:
GHS07
:
Refrigerator