tert-Butyl-4-(3-aminobenzyl)piperazine-1-carboxylate 97%
Catalog # APOSOR1382-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:291.4 g/mol
Melting Pt:107…108 °C
Storage Temperature:Ambient
UN:1759
ADR:8,III
:GHS05
:GHS07
CAS Number:361345-40-6
Formula:C₁₆H₂₅N₃O₂
MDL Number:MFCD06658979