Order Entry
Germany
ContactUsLinkComponent
tert-Butyl-4-formylpiperidine-1-carboxylate 97%
tert-Butyl-4-formylpiperidine-1-carboxylate 97%
Catalog # APOSOR13630-10G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl-4-formylpiperidine-1-carboxylate 97%
Catalog # APOSOR13630-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :213.28 g/mol
Melting Pt:34…40 °C
Flash Pt:>110 °C
 :GHS07
Storage Temperature:Freezer
CAS Number:137076-22-3
Formula:C₁₁H₁₉NO₃
MDL Number:MFCD02179019