

2-Methoxypyridine 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1628-89-3
:
Formula:C₆H₇NO
MDL Number:MFCD00006262
:109.13 g/mol
Boiling Pt:142…143 °C
Density:1.049 g/cm³ (25 °C)
Flash Pt:32 °C
Storage Temperature:Ambient
EINECS:216-623-8
UN:1993
ADR:3,III
:GHS02
:GHS07