0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
2-Mercapto-5-methoxyimidazole[4,5-b]pyridine
2-Mercapto-5-methoxyimidazole[4,5-b]pyridine
2-Mercapto-5-methoxyimidazole[4,5-b]pyridine
Irritant
2-Mercapto-5-methoxyimidazole[4,5-b]pyridine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Mercapto-5-methoxyimidazole[4,5-b]pyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
113713-60-3
:
Formula:
C₇H₇N₃OS
MDL Number:
MFCD06658230
:
181.22 g/mol
Melting Pt:
238…241 °C
:
GHS07
Storage Temperature:
Refrigerator