Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Methyl-1H-pyrazol-4-carbaldehyde
1-Methyl-1H-pyrazol-4-carbaldehyde
1-Methyl-1H-pyrazol-4-carbaldehyde
Irritant
1-Methyl-1H-pyrazol-4-carbaldehyde
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1-Methyl-1H-pyrazol-4-carbaldehyde
Supplier:
Apollo Scientific
About this item
CAS Number:
25016-11-9
:
Formula:
C₅H₆N₂O
MDL Number:
MFCD00460465
:
110.12 g/mol
Melting Pt:
29…34 °C
Flash Pt:
>110 °C
:
GHS07
Storage Temperature:
Ambient