tert-Butyl-1,4-diazepane-1-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:112275-50-0
:
Formula:C₁₀H₂₀N₂O₂
MDL Number:MFCD00276987
:200.28 g/mol
Boiling Pt:95…110 °C/0.5mmHg
Density:1.016 g/cm³ (25 °C)
Flash Pt:110 °C
:GHS07
Storage Temperature:Ambient