Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Methyl-1H-pyrazole-3-carbonitrile
1-Methyl-1H-pyrazole-3-carbonitrile 97%
1-Methyl-1H-pyrazole-3-carbonitrile 97%
APOSOR12256-100MG
:
Apollo Scientific
:
1-Methyl-1H-pyrazole-3-carbonitrile 97%
APOSOR12256-100MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 mg
Seq:
2440
:
:
107.12 g/mol
:
41.5…45 °C
:
Ambient
:
3439
:
6.1,III
:
GHS07
:
79080-39-0
:
C₅H₅N₃
:
MFCD00466343