
Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate
Catalog # APOSOR12149-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:207.185 g/mol
Melting Pt:183…185 °C
:GHS07
Storage Temperature:Ambient
CAS Number:202195-67-3
Formula:C₁₀H₉NO₄
MDL Number:MFCD03036646