
Methyl-4-methyl-2-(methylsulphanyl)-5-pyrimidinecarboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:166392-24-1
:
Formula:C₈H₁₀N₂O₂S
MDL Number:MFCD04125151
:198.25 g/mol
Melting Pt:59…61 °C
:GHS07
Storage Temperature:Ambient