
Ethyl-4-[2-(4-hydroxypiperidino)ethoxy]benzenecarboxylate
Catalog # APOSOR12097-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:293.37 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:937601-92-8
Formula:C₁₆H₂₃NO₄
MDL Number:MFCD08689771