
7-Bromo-2,3-dihydro-1-benzofuran-5-carbaldehyde
Catalog # APOSOR12038-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:227.06 g/mol
Melting Pt:109…111 °C
:GHS07
Storage Temperature:Ambient
CAS Number:875551-14-7
Formula:C₉H₇BrO₂
MDL Number:MFCD08689769