Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-Butyl 5-bromo-2-chlorobenzoate
tert-Butyl-5-bromo-2-chlorobenzoate
tert-Butyl-5-bromo-2-chlorobenzoate
Irritant
tert-Butyl-5-bromo-2-chlorobenzoate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
tert-Butyl-5-bromo-2-chlorobenzoate
Supplier:
Apollo Scientific
About this item
CAS Number:
503555-23-5
:
Formula:
C₁₁H₁₂BrClO₂
MDL Number:
MFCD09800903
:
GHS07
Storage Temperature:
Ambient