
5-Methyl-2-nitroanisole 98%
Catalog # APOSOR11561-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:167.16 g/mol
Melting Pt:58…60 °C
:GHS07
Storage Temperature:Ambient
CAS Number:38512-82-2
Formula:C₈H₉NO₃
MDL Number:MFCD00007367