Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Benzylaminopurine
6-Benzylaminopurine
6-Benzylaminopurine
6-Benzylaminopurine
Supplier:
Apollo Scientific
Certificates
Expand image
6-Benzylaminopurine
Supplier:
Apollo Scientific
Certificates
About this item
CAS Number:
1214-39-7
:
Formula:
C₁₂H₁₁N₅
MDL Number:
MFCD00005572
:
225.25 g/mol
Melting Pt:
229…233 °C
EINECS:
214-927-5
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
Documentation