
2-Methyl-5-methoxybenzimidazole
Catalog # APOSOR11211-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:GHS07
Storage Temperature:Ambient
CAS Number:4887-81-4
Formula:C₉H₁₀N₂O