Order Entry
Germany
Orders LinkContactUsLinkComponent
 

5'-Bromo-2'-hydroxyacetophenone

Bottle, 1 g
Catalog # APOSOR1113-1G
Irritant
Supplier:   Apollo Scientific
5'-Bromo-2'-hydroxyacetophenone
Catalog # APOSOR1113-1G
Supplier:  Apollo Scientific
CAS Number:  1450-75-5
5'-Bromo-2'-hydroxyacetophenone

Bottle, 1 g

CAS Number:  1450-75-5
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  1-(5-Bromo-2-hydroxyphenyl)ethan-1-one, 1-(5-Bromo-2-hydroxyphenyl)ethanone
  • CAS Number:1450-75-5
  • Formula:C₈H₇BrO₂
  • MDL Number:MFCD00191850
  • Molecular Weight:215.05
  • Melting Pt:55…61 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications