Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Methoxy-1-methyl-1H-indole-3-carbaldehyde
5-Methoxy-1-methyl-1H-indole-3-carbaldehyde
5-Methoxy-1-methyl-1H-indole-3-carbaldehyde
Irritant
5-Methoxy-1-methyl-1H-indole-3-carbaldehyde
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Methoxy-1-methyl-1H-indole-3-carbaldehyde
Supplier:
Apollo Scientific
About this item
CAS Number:
39974-94-2
:
Formula:
C₁₁H₁₁NO₂
MDL Number:
MFCD01578324
Melting Pt:
126…129 °C
:
189.214 g/mol
:
GHS07
Storage Temperature:
Refrigerator