Order Entry
Germany
ContactUsLinkComponent
3-Amino-3-(3-bromophenyl)propionic acid
Irritant
3-Amino-3-(3-bromophenyl)propionic acid
Catalog # APOSOR110160-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Amino-3-(3-bromophenyl)propionic acid
Catalog # APOSOR110160-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :244.0852 g/mol
Melting Pt:218…220 °C (dec.)
 :GHS07
Storage Temperature:Ambient
CAS Number:117391-50-1
Formula:C₉H₁₀BrNO₂
MDL Number:MFCD00995843