0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Methyl 6-bromoimidazo[1,2-a]pyridine-3-carboxylate
Methyl-6-bromoimidazo[1,2-a]pyridine-3-carboxylate
Methyl-6-bromoimidazo[1,2-a]pyridine-3-carboxylate
:
Apollo Scientific
Methyl-6-bromoimidazo[1,2-a]pyridine-3-carboxylate
:
Apollo Scientific
:
1359656-01-1
:
:
C₉H₇BrN₂O₂
:
MFCD21364589
:
GHS07
:
Ambient