Order Entry
Germany
ContactUsLinkComponent
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I)
Irritant
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I)
Catalog # APOSOR10921-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I)
Catalog # APOSOR10921-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :360.41 g/mol
Boiling Pt:380…384 °C/679mmHg
Melting Pt:160…164 °C
EINECS:238-041-3
 :GHS07
Storage Temperature:Ambient
CAS Number:14187-32-7
Formula:C₂₀H₂₄O₆
MDL Number:MFCD00005098