
2-Bromo-5-chloroaniline 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:823-57-4
:
Formula:C₆H₅BrClN
MDL Number:MFCD00672942
:206.47 g/mol
Boiling Pt:128 °C/6.75mmHg
Melting Pt:38 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together