Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Nitrophenylboronic acid
2-Nitrophenylboronic acid 97%
2-Nitrophenylboronic acid 97%
:
Apollo Scientific
2-Nitrophenylboronic acid 97%
:
Apollo Scientific
:
5570-19-4
:
:
C₆H₆BNO₄
:
MFCD00161358
:
166.93 g/mol
:
155…160 °C
:
GHS07
:
Refrigerator