Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl oxindole-5-carboxylate
Methyl oxindole-5-carboxylate 96%
Methyl oxindole-5-carboxylate 96%
Irritant
Methyl oxindole-5-carboxylate 96%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl oxindole-5-carboxylate 96%
Supplier:
Apollo Scientific
About this item
CAS Number:
199328-10-4
:
Formula:
C₁₀H₉NO₃
MDL Number:
MFCD02179600
:
191.1834 g/mol
Melting Pt:
192…199 °C
:
GHS07
Storage Temperature:
Refrigerator