Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Aminophenylboronic acid pinacol ester
2-Aminophenylboronic acid pinacol ester
2-Aminophenylboronic acid pinacol ester
2-Aminophenylboronic acid pinacol ester
APOSOR10339-25G
:
Apollo Scientific
:
2-Aminophenylboronic acid pinacol ester
APOSOR10339-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
219.09 g/mol
:
68…70 °C
:
GHS07
:
Refrigerator
:
191171-55-8
:
C₁₂H₁₈BNO₂
:
MFCD02179448