Order Entry
Germany
Orders LinkContactUsLinkComponent
2-Methylindoline

2-Methylindoline

Irritant
Supplier:   Apollo Scientific
2-Methylindoline
Supplier:  Apollo Scientific
Certificates
2-Methylindoline

2 Options Available Below

Product Details & Documents
  • CAS Number:6872-06-6
  • Formula:C₉H₁₁N
  • MDL Number:MFCD00005706
  • Molecular Weight:133.19 g/mol
  • Boiling Pt:228…229 °C
  • Melting Pt:–51 °C
  • Flash Pt:93 °C
  • Storage Temperature:Ambient
  • EINECS:229-971-0
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 21,80 €
    21,80 €
    1 * 25 g
       21,80 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 34,80 €
    34,80 €
    1 * 100 g
       34,80 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.