Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Benzofuran-2-carboxaldehyde
Benzofuran-2-carboxaldehyde
Benzofuran-2-carboxaldehyde
APOSOR101115-1G
:
Apollo Scientific
:
Benzofuran-2-carboxaldehyde
APOSOR101115-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
146.145 g/mol
:
135 °C/18mmHg
:
1.206 g/cm³ (25 °C)
:
224-248-6
:
GHS07
:
Ambient
:
4265-16-1
:
C₉H₆O₂
:
MFCD00015463