Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
trans-4-Aminocyclohexanol hydrochloride
trans-4-Aminocyclohexanol hydrochloride
trans-4-Aminocyclohexanol hydrochloride
Irritant
trans-4-Aminocyclohexanol hydrochloride
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
trans-4-Aminocyclohexanol hydrochloride
Supplier:
Apollo Scientific
About this item
CAS Number:
50910-54-8
:
Formula:
C₆H₁₄ClNO
MDL Number:
MFCD00012566
:
151.64 g/mol
Melting Pt:
225…227 °C
:
GHS07
Storage Temperature:
Ambient