
3-Chloropropyldimethylammonium chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5407-04-5
:
Formula:C₅H₁₃Cl₂N
MDL Number:MFCD00012521
:158.07 g/mol
Melting Pt:138…144 °C
EINECS:226-467-2
:GHS07
Storage Temperature:Ambient