Order Entry
Germany
ContactUsLinkComponent
DL-Tryptophan methyl ester hydrochloride 97%
DL-Tryptophan methyl ester hydrochloride 97%
Catalog # APOSOR0765-25G
Supplier:  Apollo Scientific
CAS Number:  
DL-Tryptophan methyl ester hydrochloride 97%
Catalog # APOSOR0765-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :254.714 g/mol
Melting Pt:221…222 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:5619-09-0
Formula:C₁₂H₁₅ClN₂O₂
MDL Number:MFCD00066133