Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Dimethylamino)-2-oxoacetic acid
2-(Dimethylamino)-2-oxoacetic acid
2-(Dimethylamino)-2-oxoacetic acid
Irritant
2-(Dimethylamino)-2-oxoacetic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-(Dimethylamino)-2-oxoacetic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
32833-96-8
:
Formula:
C₄H₇NO₃
MDL Number:
MFCD01971023
Melting Pt:
126…128 °C (dec.)
:
117.1000 g/mol
:
GHS07
Storage Temperature:
Ambient