Triisopropylchlorosilane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:13154-24-0
:
Formula:C₉H₂₁ClSi
MDL Number:MFCD00009656
:192.8 g/mol
Boiling Pt:79…80 °C/10mmHg
Density:0.902 g/cm³ (25 °C)
Flash Pt:62 °C
Storage Temperature:Ambient
UN:2987
ADR:8,II
:GHS05